Product Details

SNP ID
rs6702407
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:169854529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCTTGTGATGACACTTTTGGC[A/G]GCTCGATTTGGTCTGCGTCATCCCC
Phenotype
MIM: 608192
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf112 PubMed Links

Gene Details

Gene
C1orf112
Gene Name
chromosome 1 open reading frame 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320047.1 2081 Intron NP_001306976.1
NM_001320048.1 2081 Intron NP_001306977.1
NM_001320050.1 2081 Intron NP_001306979.1
NM_001320051.1 2081 Intron NP_001306980.1
NM_018186.3 2081 Intron NP_060656.2
XM_005245317.4 2081 Intron XP_005245374.1
XM_011509735.2 2081 Intron XP_011508037.1
XM_017001722.1 2081 Intron XP_016857211.1
XM_017001723.1 2081 Intron XP_016857212.1
XM_017001724.1 2081 Intron XP_016857213.1
XM_017001725.1 2081 Intron XP_016857214.1
Gene
SCYL3
Gene Name
SCY1 like pseudokinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020423.6 2081 Missense Mutation NP_065156.5
NM_181093.3 2081 Missense Mutation NP_851607.2
XM_006711465.1 2081 Missense Mutation XP_006711528.1
XM_011509801.1 2081 Missense Mutation XP_011508103.1
XM_011509802.1 2081 Missense Mutation XP_011508104.1
XM_011509803.1 2081 Missense Mutation XP_011508105.1
XM_017001862.1 2081 Missense Mutation XP_016857351.1
XM_017001863.1 2081 Missense Mutation XP_016857352.1
XM_017001864.1 2081 Missense Mutation XP_016857353.1
XM_017001865.1 2081 Missense Mutation XP_016857354.1

View Full Product Details