Product Details

SNP ID
rs7845046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:48013704 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATGCCTGGAAAATGTCTTTGAGT[A/T]ATGAGAAACTCAGAAAAAATTGATG
Phenotype
MIM: 603001
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2V2 PubMed Links

Gene Details

Gene
UBE2V2
Gene Name
ubiquitin conjugating enzyme E2 V2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003350.2 Intron NP_003341.1
XM_005251300.2 Intron XP_005251357.1
XM_011517583.2 Intron XP_011515885.1
XM_017013808.1 Intron XP_016869297.1

View Full Product Details