Product Details

SNP ID
rs10401216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57861328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAACTTTTGAGTCAGTGGTTTAT[G/T]TTTTGCATTTTTTTTGTTTTGAGAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2CP5 PubMed Links

Gene Details

Gene
UBE2CP5
Gene Name
ubiquitin conjugating enzyme E2 C pseudogene 5
There are no transcripts associated with this gene.

Gene
ZNF587
Gene Name
zinc finger protein 587
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204817.1 Intron NP_001191746.1
NM_032828.3 Intron NP_116217.1
Gene
ZNF814
Gene Name
zinc finger protein 814
There are no transcripts associated with this gene.

View Full Product Details