Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_181707.2 | Intron | NP_859058.2 | |||
XM_005257033.4 | Intron | XP_005257090.1 | |||
XM_005257034.4 | Intron | XP_005257091.1 | |||
XM_005257035.4 | Intron | XP_005257092.1 | |||
XM_005257036.2 | Intron | XP_005257093.1 | |||
XM_017024166.1 | Intron | XP_016879655.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_032582.3 | Intron | NP_115971.2 | |||
XM_011525371.1 | Intron | XP_011523673.1 | |||
XM_011525372.1 | Intron | XP_011523674.1 | |||
XM_011525373.1 | Intron | XP_011523675.1 | |||
XM_011525374.1 | Intron | XP_011523676.1 | |||
XM_011525375.1 | Intron | XP_011523677.1 | |||
XM_011525376.1 | Intron | XP_011523678.1 | |||
XM_011525378.1 | Intron | XP_011523680.1 | |||
XM_011525379.2 | Intron | XP_011523681.1 | |||
XM_017025233.1 | Intron | XP_016880722.1 | |||
XM_017025234.1 | Intron | XP_016880723.1 |