Product Details

SNP ID
rs6124685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45092878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCACTTTCTCGGGCCATGGTCC[C/T]TGATACATTATACACACCTGTACTT
Phenotype
MIM: 602905
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNS1 PubMed Links

Gene Details

Gene
KCNS1
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322799.1 3970 UTR 3 NP_001309728.1
NM_002251.4 3970 UTR 3 NP_002242.2

View Full Product Details