Product Details

SNP ID
rs9487073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109366933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTATGTTTTCACAAAATTTGCTAC[A/G]TATGTTGACATGAATGTGTGTCAGG
Phenotype
MIM: 603356
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD164 PubMed Links

Gene Details

Gene
CD164
Gene Name
CD164 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142401.1 2097 UTR 3 NP_001135873.1
NM_001142402.1 2097 UTR 3 NP_001135874.1
NM_001142403.1 2097 UTR 3 NP_001135875.1
NM_001142404.1 2097 UTR 3 NP_001135876.1
NM_006016.4 2097 UTR 3 NP_006007.2

View Full Product Details