Product Details

SNP ID
rs5942873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:110005307 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGCAGGTCATTTGCTTGCAGCCA[A/G]AAGATGAATTAGTGCCTGGGTTAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM164 PubMed Links

Gene Details

Gene
TMEM164
Gene Name
transmembrane protein 164
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017698.2 Intron NP_060168.2
NM_032227.3 Intron NP_115603.2
XM_005262205.4 Intron XP_005262262.1
XM_005262206.3 Intron XP_005262263.1
XM_005262208.4 Intron XP_005262265.1
XM_011531051.2 Intron XP_011529353.1
XM_011531054.1 Intron XP_011529356.1
XM_011531055.2 Intron XP_011529357.1
XM_017029894.1 Intron XP_016885383.1
XM_017029895.1 Intron XP_016885384.1
XM_017029896.1 Intron XP_016885385.1
XM_017029897.1 Intron XP_016885386.1
XM_017029898.1 Intron XP_016885387.1
XM_017029899.1 Intron XP_016885388.1
XM_017029900.1 Intron XP_016885389.1

View Full Product Details