Product Details
- SNP ID
-
rs5750259
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:36468360 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTTGGCCGGGTCACAGCACCTTT[C/G]TGAGCCTCAGTGTCCTTTTCCATAA
- Phenotype
-
MIM: 609063
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TXN2
PubMed Links
Gene Details
- Gene
- TXN2
- Gene Name
- thioredoxin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012473.3 |
|
Intron |
|
|
NP_036605.2 |
View Full Product Details