Product Details

SNP ID
rs9655959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:88279073 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATACTTCTATTGGATTTTGCAAGA[C/T]ATTAGGTCATGCATGTTGCCCATAA
Phenotype
MIM: 611098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STEAP4 PubMed Links

Gene Details

Gene
STEAP4
Gene Name
STEAP4 metalloreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205315.1 1928 UTR 3 NP_001192244.1
NM_001205316.1 1928 UTR 3 NP_001192245.1
NM_024636.3 1928 UTR 3 NP_078912.2

View Full Product Details