Product Details

SNP ID
rs9941591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233476052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACGGAAACGCACAGACGTCTAT[A/G]GCAGACGCTCTCAGACACGTGTGCA
Phenotype
MIM: 601826 MIM: 610570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKD PubMed Links

Gene Details

Gene
DGKD
Gene Name
diacylglycerol kinase delta
There are no transcripts associated with this gene.

Gene
USP40
Gene Name
ubiquitin specific peptidase 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018218.2 5176 UTR 3 NP_060688.1
XM_006712612.3 5176 UTR 3 XP_006712675.1
XM_011511396.2 5176 UTR 3 XP_011509698.1
XM_011511397.2 5176 UTR 3 XP_011509699.1
XM_011511398.2 5176 Intron XP_011509700.1
XM_011511399.2 5176 UTR 3 XP_011509701.1
XM_011511400.2 5176 UTR 3 XP_011509702.1
XM_011511401.2 5176 Intron XP_011509703.1
XM_017004426.1 5176 UTR 3 XP_016859915.1
XM_017004427.1 5176 UTR 3 XP_016859916.1
XM_017004428.1 5176 UTR 3 XP_016859917.1

View Full Product Details