Product Details

SNP ID
rs9982493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:31877035 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGTACTCCCAGCCCACGTGGAG[C/T]GAAAGGATTCAGGGCAGCTTGTCTT
Phenotype
MIM: 606532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HUNK PubMed Links

Gene Details

Gene
HUNK
Gene Name
hormonally up-regulated Neu-associated kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014586.1 Intron NP_055401.1
XM_011529537.2 Intron XP_011527839.1

View Full Product Details