Product Details

SNP ID
rs10494836
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:202727750 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGAGGCAGAGTCTGGGAATTCA[C/T]AAGTCTCCTCTCAAGACATATACTA
Phenotype
MIM: 605393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM5B PubMed Links

Gene Details

Gene
KDM5B
Gene Name
lysine demethylase 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314042.1 7194 UTR 3 NP_001300971.1
NM_006618.4 7194 UTR 3 NP_006609.3
XM_011509088.2 7194 UTR 3 XP_011507390.1
XM_011509090.2 7194 UTR 3 XP_011507392.1
XM_011509091.2 7194 UTR 3 XP_011507393.1
XM_011509092.2 7194 UTR 3 XP_011507394.1

View Full Product Details