Product Details

SNP ID
rs9854618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187218990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCACCTCATGCAGCCCAGTTT[C/T]CCAAGAAGGGAAGGAGGCCAGGGCC
Phenotype
MIM: 600521
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MASP1 PubMed Links

Gene Details

Gene
MASP1
Gene Name
mannan binding lectin serine peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031849.2 3571 Intron NP_001027019.1
NM_001879.5 3571 UTR 3 NP_001870.3
NM_139125.3 3571 Intron NP_624302.1
XM_006713701.2 3571 Intron XP_006713764.1
XM_011512989.2 3571 Intron XP_011511291.1
XM_011512990.2 3571 Intron XP_011511292.1
XM_011512991.2 3571 Intron XP_011511293.1
XM_017006869.1 3571 Intron XP_016862358.1
XM_017006870.1 3571 Intron XP_016862359.1
XM_017006871.1 3571 Intron XP_016862360.1
XM_017006872.1 3571 Intron XP_016862361.1

View Full Product Details