Product Details

SNP ID
rs5962279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:104167904 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACATATACAAAGGGCTGGGGGGG[G/T]GTGTGGTTAAGGCTCTAGGAGAGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM199X PubMed Links

Gene Details

Gene
FAM199X
Gene Name
family with sequence similarity 199, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207318.3 Intron NP_997201.1
XM_005262079.2 Intron XP_005262136.1
XM_017029270.1 Intron XP_016884759.1

View Full Product Details