Product Details

SNP ID
rs6696571
Assay Type
Functionally Tested
NCBI dbSNP Submissions
20
Location
Chr.1:240780472 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAAAAAAAAAAAAAAGTGCTT[C/T]CTCTTTGTGGGATGAAGGAAGGTAC
Phenotype
MIM: 602517
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RGS7 PubMed Links

Gene Details

Gene
RGS7
Gene Name
regulator of G-protein signaling 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282773.1 Intron NP_001269702.1
NM_001282775.1 Intron NP_001269704.1
NM_001282778.1 Intron NP_001269707.1
NM_002924.5 Intron NP_002915.3
XM_005273218.3 Intron XP_005273275.1
XM_006711800.3 Intron XP_006711863.1
XM_011544246.2 Intron XP_011542548.1
XM_011544247.2 Intron XP_011542549.1
XM_017002001.1 Intron XP_016857490.1
XM_017002002.1 Intron XP_016857491.1
XM_017002003.1 Intron XP_016857492.1
XM_017002004.1 Intron XP_016857493.1
XM_017002005.1 Intron XP_016857494.1
XM_017002006.1 Intron XP_016857495.1
XM_017002007.1 Intron XP_016857496.1
XM_017002008.1 Intron XP_016857497.1
XM_017002009.1 Intron XP_016857498.1
XM_017002010.1 Intron XP_016857499.1
XM_017002011.1 Intron XP_016857500.1
XM_017002012.1 Intron XP_016857501.1
XM_017002013.1 Intron XP_016857502.1

View Full Product Details