Product Details

SNP ID
rs5918922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:65078310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAAATTGTAAATACAGCTGTGA[C/G]ACTCAGTACAATGACTTTAGTGAAT
Phenotype
MIM: 300889
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZC3H12B PubMed Links

Gene Details

Gene
ZC3H12B
Gene Name
zinc finger CCCH-type containing 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010888.3 Intron NP_001010888.3

View Full Product Details