Product Details
- SNP ID
-
hCV29565292
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:55851541 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTATGAAAATTGAGACAGGAAGGCA[A/G]AGTGTCAGCTTGTTCCACCTCAGCT
- Phenotype
-
MIM: 609510
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL31RA
PubMed Links
Gene Details
- Gene
- IL31RA
- Gene Name
- interleukin 31 receptor A
There are no transcripts associated with this gene.
View Full Product Details