Product Details
- SNP ID
-
rs6103330
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:43458814 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAGGGGTGGCTGTATTTGTTGTAT[C/T]CTCCTTACTTACCTATCAGTGCATT
- Phenotype
-
MIM: 601944
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SRSF6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112824898,rs6103331] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SRSF6
- Gene Name
- serine and arginine rich splicing factor 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006275.5 |
|
Intron |
|
|
NP_006266.2 |
View Full Product Details