Product Details

SNP ID
rs6142116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34187567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTTAGGAGGATCGTGAATTTC[C/T]TAAACTGTCTTTGAGAGACTCAGAT
Phenotype
MIM: 600201
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASIP PubMed Links

Gene Details

Gene
ASIP
Gene Name
agouti signaling protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001672.2 Intron NP_001663.2
XM_005260412.3 Intron XP_005260469.1
XM_011528820.2 Intron XP_011527122.1
XM_011528821.1 Intron XP_011527123.1

View Full Product Details