Product Details

SNP ID
rs9332701
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:169515529 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGATGGTATAGCTCTTTACATAC[A/G]TTTCAGAGGACAGAGACTTGCAGCC
Phenotype
MIM: 612309
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F5 PubMed Links

Gene Details

Gene
F5
Gene Name
coagulation factor V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000130.4 6588 Missense Mutation ACG,ATG T2148M NP_000121.2
XM_017000660.1 6588 Missense Mutation ACG,ATG T2011M XP_016856149.1

View Full Product Details