Product Details

SNP ID
rs4796406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7340341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTTGTTGCACATATCAACAGTT[C/T]GTTCCTTTTTATTGCTGAGTACTGT
Phenotype
MIM: 607763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAP1 PubMed Links

Gene Details

Gene
ACAP1
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014716.3 Intron NP_055531.1

View Full Product Details