Product Details

SNP ID
rs4986948
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.11:67585234 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TGCAAATACATCTCCCTCATCTACA[C/G]CAACTATGTGAGCATCTGCACCAGG
Phenotype
MIM: 134660
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GSTP1 PubMed Links

Gene Details

Gene
GSTP1
Gene Name
glutathione S-transferase pi 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000852.3 578 Missense Mutation ACC,AGC T110S NP_000843.1

View Full Product Details