Product Details

SNP ID
rs6004297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24813813 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTGTGTCTCTTGTACCATATGGC[C/T]GGGTCTCCCCCAGCCACCTCATGAG
Phenotype
MIM: 611417
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGSM1 PubMed Links

Gene Details

Gene
SGSM1
Gene Name
small G protein signaling modulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039948.3 Intron NP_001035037.1
NM_001098497.2 Intron NP_001091967.1
NM_001098498.2 Intron NP_001091968.1
NM_133454.3 Intron NP_597711.1

View Full Product Details