Product Details

SNP ID
rs9916685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75827639 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGAGGCAGATGGGCCAGGGCCAG[A/G]AGACAGATGGCCCAATCCCCTGCCC
Phenotype
MIM: 608897 MIM: 616375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100507191 PubMed Links

Gene Details

Gene
LOC100507191
Gene Name
uncharacterized LOC100507191
There are no transcripts associated with this gene.

Gene
UNC13D
Gene Name
unc-13 homolog D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199242.2 3678 UTR 3 NP_954712.1
XM_011524504.1 3678 UTR 3 XP_011522806.1
XM_011524506.1 3678 UTR 3 XP_011522808.1
XM_011524507.2 3678 UTR 3 XP_011522809.1
Gene
UNK
Gene Name
unkempt family zinc finger
There are no transcripts associated with this gene.

View Full Product Details