Product Details

SNP ID
rs9940279
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70590236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTCCTAGCATGTGTGGGATGGG[C/G]AGATTCCAGACATTGGAAAGTCCCT
Phenotype
MIM: 612081
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
IL34 PubMed Links

Gene Details

Gene
IL34
Gene Name
interleukin 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172771.1 75 Intron NP_001166242.1
NM_001172772.1 75 Intron NP_001166243.1
NM_152456.2 75 Intron NP_689669.2
XM_011522902.2 75 UTR 5 XP_011521204.1
XM_011522903.2 75 Intron XP_011521205.1

View Full Product Details