Product Details

SNP ID
rs10271886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1684961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTCTGGTACTCTTCATTTCTTT[C/T]TGTGAATTTGAATTACCATCTGACA
Phenotype
MIM: 614964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELFN1 PubMed Links

Gene Details

Gene
ELFN1
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128636.2 Intron NP_001122108.1
XM_006715725.3 Intron XP_006715788.1
XM_006715726.2 Intron XP_006715789.1
XM_006715727.3 Intron XP_006715790.1
XM_011515397.2 Intron XP_011513699.1
XM_011515398.2 Intron XP_011513700.1
XM_011515401.2 Intron XP_011513703.1
XM_017012203.1 Intron XP_016867692.1
Gene
LOC401296
Gene Name
uncharacterized LOC401296
There are no transcripts associated with this gene.

View Full Product Details