Product Details

SNP ID
rs10105278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:26742551 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGCCCAGAGCAGAGATACAAATT[A/C]CTCTAAATTAATGTATGAATCTGGC
Phenotype
MIM: 104221
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADRA1A PubMed Links

Gene Details

Gene
ADRA1A
Gene Name
adrenoceptor alpha 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000680.3 12071 Intron NP_000671.2
NM_001322502.1 12071 Intron NP_001309431.1
NM_001322503.1 12071 Intron NP_001309432.1
NM_001322504.1 12071 Intron NP_001309433.1
NM_033302.3 12071 Intron NP_150645.2
NM_033303.4 12071 Intron NP_150646.3
NM_033304.3 12071 Intron NP_150647.2
XM_006716292.3 12071 UTR 3 XP_006716355.1
XM_006716293.3 12071 Intron XP_006716356.1
XM_011544411.2 12071 Intron XP_011542713.1
XM_011544412.2 12071 Intron XP_011542714.1
XM_017013094.1 12071 UTR 3 XP_016868583.1
XM_017013095.1 12071 UTR 3 XP_016868584.1
XM_017013096.1 12071 Intron XP_016868585.1

View Full Product Details