Product Details

SNP ID
rs5760005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23762646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGACCTGGAAGCGACTAGGGCTGA[G/T]TACAAAGGGGGCAAAGGAATTTGGG
Phenotype
MIM: 615903 MIM: 605017
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C22orf15 PubMed Links

Gene Details

Gene
C22orf15
Gene Name
chromosome 22 open reading frame 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182520.2 123 Intron NP_872326.2
XM_011529907.2 123 Intron XP_011528209.1
XM_011529908.2 123 UTR 5 XP_011528210.2
XM_011529912.2 123 Intron XP_011528214.1
XM_017028602.1 123 Intron XP_016884091.1
XM_017028603.1 123 Intron XP_016884092.1
XM_017028604.1 123 Intron XP_016884093.1
XM_017028605.1 123 Intron XP_016884094.1
XM_017028606.1 123 Intron XP_016884095.1
XM_017028607.1 123 Intron XP_016884096.1
XM_017028608.1 123 Intron XP_016884097.1
XM_017028609.1 123 UTR 5 XP_016884098.1
XM_017028610.1 123 UTR 5 XP_016884099.1
XM_017028611.1 123 Intron XP_016884100.1
XM_017028612.1 123 UTR 5 XP_016884101.1
XM_017028613.1 123 Intron XP_016884102.1
XM_017028614.1 123 Intron XP_016884103.1
XM_017028615.1 123 UTR 5 XP_016884104.1
XM_017028616.1 123 UTR 5 XP_016884105.1
XM_017028617.1 123 UTR 5 XP_016884106.1
XM_017028618.1 123 Intron XP_016884107.1
Gene
CHCHD10
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
There are no transcripts associated with this gene.

Gene
VPREB3
Gene Name
pre-B lymphocyte 3
There are no transcripts associated with this gene.

View Full Product Details