Product Details

SNP ID
rs10482481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:115289930 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAGACCTCGGCATAGCGCTTACT[A/G]GAAGTGGCGTCGGGAGGAACACACC
Phenotype
MIM: 300616
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LUZP4 PubMed Links

Gene Details

Gene
LUZP4
Gene Name
leucine zipper protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318840.1 Intron NP_001305769.1
NM_016383.4 Intron NP_057467.1

View Full Product Details