Product Details

SNP ID
rs1078699
Assay Type
Functionally tested
NCBI dbSNP Submissions
54
Location
Chr.1:155265702 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAATTTAAGTCTTCTTAAGAGAT[C/T]TCTCCCTTTCAACCTATTTAGTTTT
Phenotype
MIM: 602989 MIM: 606913
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLK2 PubMed Links

Gene Details

Gene
CLK2
Gene Name
CDC like kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294338.1 Intron NP_001281267.1
NM_001294339.1 Intron NP_001281268.1
NM_003993.3 Intron NP_003984.2
XM_005244878.1 Intron XP_005244935.1
XM_011509143.1 Intron XP_011507445.1
XM_011509144.1 Intron XP_011507446.1
XM_011509147.2 Intron XP_011507449.1
XM_017000241.1 Intron XP_016855730.1
XM_017000242.1 Intron XP_016855731.1
XM_017000243.1 Intron XP_016855732.1
XM_017000244.1 Intron XP_016855733.1
Gene
SCAMP3
Gene Name
secretory carrier membrane protein 3
There are no transcripts associated with this gene.

View Full Product Details