Product Details

SNP ID
rs5935917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15249260 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTATTATTAATGGAACCTAAGT[A/G]TTAGCCAGTACAGCCTGACGGAGAG
Phenotype
MIM: 300890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB9 PubMed Links

Gene Details

Gene
ASB9
Gene Name
ankyrin repeat and SOCS box containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031739.2 Intron NP_001026909.1
NM_001168530.1 Intron NP_001162002.1
NM_001168531.1 Intron NP_001162003.1
NM_024087.2 Intron NP_076992.1
XM_005274446.1 Intron XP_005274503.1
XM_011545458.1 Intron XP_011543760.1
XM_011545460.1 Intron XP_011543762.1
XM_017029283.1 Intron XP_016884772.1
XM_017029284.1 Intron XP_016884773.1
XM_017029285.1 Intron XP_016884774.1
XM_017029286.1 Intron XP_016884775.1

View Full Product Details