Product Details
- SNP ID
-
rs9472220
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:44162157 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAGAACTTCTGACTCAATCTCTCT[C/G]TGGAGGCCCTGGCTTCCACCCAGCT
- Phenotype
-
MIM: 604822
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CAPN11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111757201] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAPN11
- Gene Name
- calpain 11
- Gene
- TMEM63B
- Gene Name
- transmembrane protein 63B
There are no transcripts associated with this gene.
View Full Product Details