Product Details
- SNP ID
-
rs6703748
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
38
- Location
-
Chr.1:45630304 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTAGATTGTGGAAAAGATGGCTGA[A/G]TTTGAGAGAGAATGCTTGATGAGCA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78963973] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC17
- Gene Name
- coiled-coil domain containing 17
There are no transcripts associated with this gene.
- Gene
- GPBP1L1
- Gene Name
- GC-rich promoter binding protein 1 like 1
View Full Product Details