Product Details

SNP ID
rs10053046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132485866 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACACACACCCTCCCGGTGGACCT[A/G]TCTGCCATAGAGATTCCAGAACAGG
Phenotype
MIM: 147575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C5orf56 PubMed Links

Gene Details

Gene
C5orf56
Gene Name
chromosome 5 open reading frame 56
There are no transcripts associated with this gene.

Gene
IRF1
Gene Name
interferon regulatory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002198.2 Intron NP_002189.1
XM_011543378.2 Intron XP_011541680.1
XM_011543379.2 Intron XP_011541681.1

View Full Product Details