Product Details
- SNP ID
-
rs9636101
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:62045913 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTGAGTTTCTTCGTTGTGAGCAG[A/C]TGGGCCAGGCCATTGAGGAACACCA
- Phenotype
-
MIM: 606097
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PIGN
PubMed Links
Gene Details
- Gene
- PIGN
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class N
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012327.5 |
2343 |
Missense Mutation |
CAG,CAT |
Q913H |
NP_036459.1 |
NM_176787.4 |
2343 |
Missense Mutation |
CAG,CAT |
Q913H |
NP_789744.1 |
XM_011525889.1 |
2343 |
Missense Mutation |
CAG,CAT |
Q952H |
XP_011524191.1 |
XM_011525890.1 |
2343 |
Missense Mutation |
CAG,CAT |
Q952H |
XP_011524192.1 |
XM_011525891.1 |
2343 |
Missense Mutation |
CAG,CAT |
Q952H |
XP_011524193.1 |
XM_011525892.1 |
2343 |
Missense Mutation |
CAG,CAT |
Q952H |
XP_011524194.1 |
XM_011525893.1 |
2343 |
Missense Mutation |
CAG,CAT |
Q952H |
XP_011524195.1 |
XM_011525894.1 |
2343 |
Missense Mutation |
CAG,CAT |
Q952H |
XP_011524196.1 |
XM_011525895.1 |
2343 |
Missense Mutation |
CAG,CAT |
Q952H |
XP_011524197.1 |
XM_011525896.1 |
2343 |
Missense Mutation |
CAG,CAT |
Q933H |
XP_011524198.1 |
XM_011525898.1 |
2343 |
Intron |
|
|
XP_011524200.1 |
XM_017025685.1 |
2343 |
Missense Mutation |
CAG,CAT |
Q818H |
XP_016881174.1 |
XM_017025686.1 |
2343 |
Missense Mutation |
CAG,CAT |
Q779H |
XP_016881175.1 |
View Full Product Details