Product Details
- SNP ID
-
rs7544445
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
24
- Location
-
Chr.1:12750620 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGCTTTAGGTTAAAAACTATTAT[A/G]AGACAAAGAGGGACACTATATATTG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf158
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs547501039] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf158
- Gene Name
- chromosome 1 open reading frame 158
View Full Product Details