Product Details
- SNP ID
-
rs9521974
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:110882133 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCACCCTCCTATCCGAGCCCTTT[C/T]CAGGGGTGGGGACACTTGACCCTCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD10
PubMed Links
Gene Details
- Gene
- ANKRD10
- Gene Name
- ankyrin repeat domain 10
View Full Product Details