Product Details

SNP ID
rs10422202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1040745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATTAATGCTACATAAGGTGTGGG[C/G]AGAGCTGGGGTCGTGCCTCCAGCTG
Phenotype
MIM: 605414 MIM: 602373
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABCA7 PubMed Links

Gene Details

Gene
ABCA7
Gene Name
ATP binding cassette subfamily A member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019112.3 Intron NP_061985.2
XM_006722616.1 Intron XP_006722679.1
XM_006722617.2 Intron XP_006722680.1
XM_006722618.2 Intron XP_006722681.1
XM_011527628.2 Intron XP_011525930.1
XM_011527629.1 Intron XP_011525931.1
XM_011527630.1 Intron XP_011525932.1
XM_011527631.1 Intron XP_011525933.1
XM_011527632.1 Intron XP_011525934.1
XM_011527633.2 Intron XP_011525935.1
XM_011527634.1 Intron XP_011525936.1
XM_011527635.1 Intron XP_011525937.1
XM_011527636.2 Intron XP_011525938.1
XM_017026142.1 Intron XP_016881631.1
XM_017026143.1 Intron XP_016881632.1
Gene
CNN2
Gene Name
calponin 2
There are no transcripts associated with this gene.

View Full Product Details