Product Details

SNP ID
rs9494119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:134918225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTACTTATAAGTCTTTTTTTCCGA[A/G]TCCACATTGAAAATAGACAGTATCT
Phenotype
MIM: 606467
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH8A1 PubMed Links

Gene Details

Gene
ALDH8A1
Gene Name
aldehyde dehydrogenase 8 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193480.1 1573 UTR 3 NP_001180409.1
NM_022568.3 1573 UTR 3 NP_072090.1
NM_170771.2 1573 UTR 3 NP_739577.1

View Full Product Details