Product Details
- SNP ID
-
rs9377227
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:149599942 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAAAAACAAGGAGTTTCGGCATA[C/G]GTATGAAACTCAAAACAGGCCAAAT
- Phenotype
-
MIM: 606696
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GINM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75685033] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GINM1
- Gene Name
- glycoprotein integral membrane 1
There are no transcripts associated with this gene.
- Gene
- KATNA1
- Gene Name
- katanin catalytic subunit A1
- Gene
- RPS18P9
- Gene Name
- ribosomal protein S18 pseudogene 9
There are no transcripts associated with this gene.
View Full Product Details