Product Details

SNP ID
rs9901895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19649809 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGATCTGAGATTCCAAGTTTCT[A/T]ACAAGGTCCCCAGTGATATCATATG
Phenotype
MIM: 609523
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDH3A2 PubMed Links

Gene Details

Gene
ALDH3A2
Gene Name
aldehyde dehydrogenase 3 family member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000382.2 Intron NP_000373.1
NM_001031806.1 Intron NP_001026976.1
XM_011523732.1 Intron XP_011522034.1
XM_011523733.1 Intron XP_011522035.1
XM_017024355.1 Intron XP_016879844.1
XM_017024356.1 Intron XP_016879845.1
XM_017024357.1 Intron XP_016879846.1
XM_017024358.1 Intron XP_016879847.1

View Full Product Details