Product Details

SNP ID
rs4647157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:60874096 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGCCCCAAAATGTTTAACATAAA[G/T]CATGGTTAATTAAATGAAACCACTG
Phenotype
MIM: 609412
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ERCC8 PubMed Links

Gene Details

Gene
ERCC8
Gene Name
ERCC excision repair 8, CSA ubiquitin ligase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000082.3 1698 UTR 3 NP_000073.1
NM_001007233.2 1698 UTR 3 NP_001007234.1
NM_001007234.2 1698 Intron NP_001007235.1
NM_001290285.1 1698 UTR 3 NP_001277214.1

View Full Product Details