Product Details

SNP ID
rs9282771
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:58781284 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTCAACTCCACCTAGATAGCTA[A/G]TATCTACAGATGATGCAGAAAAGAG
Phenotype
MIM: 165160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JUN PubMed Links

Gene Details

Gene
JUN
Gene Name
Jun proto-oncogene, AP-1 transcription factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002228.3 2830 UTR 3 NP_002219.1
Gene
LINC01135
Gene Name
long intergenic non-protein coding RNA 1135
There are no transcripts associated with this gene.

View Full Product Details