Product Details

SNP ID
rs10512432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30485102 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGACAATAACGTGGTGGGTAAGC[A/G]TGTACTTTCAGCTAGCTTTGAAGAT
Phenotype
MIM: 604026
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOSR1 PubMed Links

Gene Details

Gene
GOSR1
Gene Name
golgi SNAP receptor complex member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007024.1 Intron NP_001007025.1
NM_001007025.1 Intron NP_001007026.1
NM_004871.2 Intron NP_004862.1
XM_005258070.4 Intron XP_005258127.1
XM_005258071.4 Intron XP_005258128.1
XM_005258072.2 Intron XP_005258129.1
XM_017025374.1 Intron XP_016880863.1
XM_017025375.1 Intron XP_016880864.1
XM_017025376.1 Intron XP_016880865.1
XM_017025377.1 Intron XP_016880866.1

View Full Product Details