Product Details
- SNP ID
-
rs9906386
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:30118751 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAAATGTGTGCTTTTCTTTTCTCT[C/T]TTTTTTTTTTTTTTTGAAACAGTGT
- Phenotype
-
MIM: 616173
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3184
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs570877718] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR3184
- Gene Name
- microRNA 3184
There are no transcripts associated with this gene.
- Gene
- MIR423
- Gene Name
- microRNA 423
There are no transcripts associated with this gene.
- Gene
- NSRP1
- Gene Name
- nuclear speckle splicing regulatory protein 1
View Full Product Details