Product Details

SNP ID
rs4145214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:152698368 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTGGGTATTAAGTACAGATTTTG[G/T]CTGGTGTTACAAAATATAATTAGGA
Phenotype
MIM: 616805
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MYCT1 PubMed Links

Gene Details

Gene
MYCT1
Gene Name
myc target 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025107.2 Intron NP_079383.2
XM_011536149.2 Intron XP_011534451.1
XM_011536150.2 Intron XP_011534452.1
XM_011536151.2 Intron XP_011534453.1
XM_011536152.2 Intron XP_011534454.1
XM_017011319.1 Intron XP_016866808.1

View Full Product Details