Product Details

SNP ID
rs9851035
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149320904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATTTAGTTTTCATCCTAAACTT[C/T]GACTATTAGTGTTTCATTTGCTTTA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TM4SF18 PubMed Links

Gene Details

Gene
TM4SF18
Gene Name
transmembrane 4 L six family member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184723.1 1270 UTR 3 NP_001171652.1
NM_138786.3 1270 UTR 3 NP_620141.1

View Full Product Details