Product Details
- SNP ID
-
rs10495940
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:46919593 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGATTACTATACATGAAATTGATC[A/C]GAAGTCTAACAAATGATTGAGGTAA
- Phenotype
-
MIM: 607788
MIM: 609332
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MCFD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116624303] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MCFD2
- Gene Name
- multiple coagulation factor deficiency 2
- Gene
- TTC7A
- Gene Name
- tetratricopeptide repeat domain 7A
View Full Product Details