Product Details

SNP ID
rs6643670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154403284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCCTACCTTGCTCTGCCAGTG[A/C]TGGGAGACCTCCAGAAACACATCGT
Phenotype
MIM: 300081 MIM: 312173 MIM: 300394
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNASE1L1 PubMed Links

Gene Details

Gene
DNASE1L1
Gene Name
deoxyribonuclease I-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009932.2 1141 Missense Mutation NP_001009932.1
NM_001009933.2 1141 Missense Mutation NP_001009933.1
NM_001009934.2 1141 Missense Mutation NP_001009934.1
NM_001303620.1 1141 Missense Mutation NP_001290549.1
NM_006730.3 1141 Missense Mutation NP_006721.1
XM_005277829.4 1141 Missense Mutation XP_005277886.1
XM_011531122.2 1141 Missense Mutation XP_011529424.1
XM_017029332.1 1141 Missense Mutation XP_016884821.1
Gene
LOC107987332
Gene Name
uncharacterized LOC107987332
There are no transcripts associated with this gene.

Gene
RPL10
Gene Name
ribosomal protein L10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256577.2 1141 Intron NP_001243506.2
NM_001256580.2 1141 Intron NP_001243509.2
NM_001303624.1 1141 Intron NP_001290553.1
NM_001303625.1 1141 Intron NP_001290554.1
NM_001303626.1 1141 Intron NP_001290555.1
NM_006013.4 1141 Intron NP_006004.3
Gene
SNORA70
Gene Name
small nucleolar RNA, H/ACA box 70
There are no transcripts associated with this gene.

Gene
TAZ
Gene Name
tafazzin
There are no transcripts associated with this gene.

View Full Product Details