Product Details

SNP ID
rs9324841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:155740441 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAATAGCAATGAAAAGCATTAT[C/T]AGTCTGGGTGGTTCTTTTTCAGATG
Phenotype
MIM: 601411
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGCD PubMed Links

Gene Details

Gene
SGCD
Gene Name
sarcoglycan delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000337.5 Intron NP_000328.2
NM_001128209.1 Intron NP_001121681.1
NM_172244.2 Intron NP_758447.1
XM_005265966.4 Intron XP_005266023.1
XM_005265967.2 Intron XP_005266024.1
XM_011534621.2 Intron XP_011532923.1
XM_017009723.1 Intron XP_016865212.1
XM_017009724.1 Intron XP_016865213.1

View Full Product Details